EMAIL THIS PAGE TO A FRIEND
European journal of immunology
Regulation of the autophagic machinery in human neutrophils.
PMID 20162553
Abstract
The induction of the autophagy machinery, a process for the catabolism of cytosolic proteins and organelles, constitutes a crucial mechanism in innate immunity. However, the involvement of autophagy in human neutrophils and the possible inducers of this process have not been completely elucidated. In this study, the induction of autophagy was examined in human neutrophils treated with various activators and detected by the formation of acidified autophagosomes through monodansylcadaverine staining and via LC-3B conversion screened by immunoblotting and immunofluorescence confocal microscopy. In addition, the expression of the ATG genes was assessed by real-time RT-PCR. We provide evidence that autophagy is implicated in human neutrophils in both a phagocytosis-independent (rapamycin, TLR agonists, PMA) and phagocytosis (Escherichia coli)-dependent initiation manner. ROS activation is a positive mechanism for autophagy induction in the case of PMA, TLR activation and phagocytosis. Furthermore, LC3B gene expression was uniformly upregulated, indicating a transcriptional level of regulation for the autophagic machinery. This study provides a stepping stone toward further investigation of autophagy in neutrophil-driven inflammatory disorders.