EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Comparison and prediction of the retention in micellar electrokinetic chromatography and microemulsion electrokinetic chromatography for disubstituted benzenes.
PMID 20162592
Abstract
Retention index (I), rather than retention factor (k), was found to be a more reasonable parameter for comparison of the relative affinity of disubstituted benzenes in MEEKC and MEKC, due to independent of I with the SDS surfactant concentration. MEKC and MEEKC may give similar or different I values, depending on types of moieties. With known I and K(ow) for alkylbenzenes as references in MEKC and MEEKC, the values of K(ow) for disubstituted benzenes can be estimated from the observed I values, where K(ow) is the octanol-water distribution constant. In addition, a group additive approach can be used to predict I for disubstituted benzenes with different moieties from the average observed I for the disubstituted benzenes with same moieties. However, electronic effects and/or intramolecular interaction may result in the different observed I from prediction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3415
Bisphenol A diglycidyl ether
C21H24O4
15138
Bisphenol A diglycidyl ether, analytical standard
C21H24O4