EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Pd-PEPPSI-IPent: low-temperature negishi cross-coupling for the preparation of highly functionalized, tetra-ortho-substituted biaryls.
PMID 20162639
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
732117
Pd-PEPPSI-IPent catalyst, ≥95%
C40H56Cl3N3Pd