EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of phalluside-1 and Sch II using 1,2-metallate rearrangements.
PMID 20165812
Abstract
(4E,8E,10E)-9-Methyl-4,8,10-sphingatrienine, a core component of marine sphingolipids, was synthesised for the first time using a copper(I)-mediated 1,2-metallate rearrangement of a lithiated glycal as a key step. It was converted to phalluside-1, a cerebroside isolated from the ascidian Phallusia fumigate. By an analogous route, (4E,8E)-9-methyl-4,8-sphingadiene was synthesised and converted to Sch II, a cerebroside that induces fruiting body formation in the basidiomycete Schizophyllum commune.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251860
1-Iodononane, 95%
C9H19I