EMAIL THIS PAGE TO A FRIEND
Parasitology research
Isolation of an antileishmanial and antitrypanosomal flavanone from the leaves of Baccharis retusa DC. (Asteraceae).
PMID 20165875
Abstract
In the course of selection of new bioactive compounds from Brazilian flora, the crude MeOH extract from the leaves of Baccharis retusa DC. (Asteraceae) showed potential against Leishmania sp. and Trypanosoma cruzi. Chromatographic fractionation of the dichloromethane phase from MeOH extract yielded great amounts of the bioactive derivative, which was characterized as 5,6,7-trihydroxy-4'-methoxyflavanone. The structure of this compound was established on the basis of spectroscopic data analysis, mainly nuclear magnetic resonance and mass spectrometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
102032
Flavanone, 98%
C15H12O2