EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Asymmetric cooperative catalysis of strong Brønsted acid-promoted reactions using chiral ureas.
PMID 20167783
Abstract
Cationic organic intermediates participate in a wide variety of useful synthetic transformations, but their high reactivity can render selectivity in competing pathways difficult to control. Here, we describe a strategy for inducing enantioselectivity in reactions of protio-iminium ions, wherein a chiral catalyst interacts with the highly reactive intermediate through a network of noncovalent interactions. This interaction leads to an attenuation of the reactivity of the iminium ion and allows high enantioselectivity in cycloadditions with electron-rich alkenes (the Povarov reaction). A detailed experimental and computational analysis of this catalyst system has revealed the precise nature of the catalyst-substrate interactions and the likely basis for enantioinduction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
225193
Sodium 3-nitrobenzenesulfonate, 98%
C6H4NNaO5S