EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Peptidomimetic antibiotics target outer-membrane biogenesis in Pseudomonas aeruginosa.
PMID 20167788
Abstract
Antibiotics with new mechanisms of action are urgently required to combat the growing health threat posed by resistant pathogenic microorganisms. We synthesized a family of peptidomimetic antibiotics based on the antimicrobial peptide protegrin I. Several rounds of optimization gave a lead compound that was active in the nanomolar range against Gram-negative Pseudomonas spp., but was largely inactive against other Gram-negative and Gram-positive bacteria. Biochemical and genetic studies showed that the peptidomimetics had a non-membrane-lytic mechanism of action and identified a homolog of the beta-barrel protein LptD (Imp/OstA), which functions in outer-membrane biogenesis, as a cellular target. The peptidomimetic showed potent antimicrobial activity in a mouse septicemia infection model. Drug-resistant strains of Pseudomonas are a serious health problem, so this family of antibiotics may have important therapeutic applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1346
ASB-14
C22H46N2O4S