EMAIL THIS PAGE TO A FRIEND
Federal register
Classification of three steroids as schedule III anabolic steroids under the Controlled Substances Act. Final rule.
PMID 20169675
Abstract
With the issuance of this final rule, the Deputy Administrator of the Drug Enforcement Administration (DEA) classifies the following three steroids as "anabolic steroids" under the Controlled Substances Act (CSA): Boldione, desoxymethyltestosterone, and 19-nor-4,9(10)-androstadienedione. These steroids and their salts, esters, and ethers are schedule III controlled substances subject to the regulatory control provisions of the CSA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7505
1,4-Androstadiene-3,17-dione
C19H24O2