EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
The use of phospholipid nanoparticles for increasing Semax resistance in various proteolytic biological media.
PMID 20170058
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8826
Semax, ≥98% (HPLC)
C37H51N9O10S · xC2HF3O2