EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
The current level of dioxin pollution in the area of large-scale spraying of Agent Orange in Vietnam.
PMID 20170064