EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
An organocatalytic asymmetric chlorolactonization.
PMID 20170118
Abstract
A reagent controlled organocatalytic enantioselective chlorolactonization reaction has been developed. Several 4-aryl pentenoic acids were cyclized in the presence of 0.1 equiv of (DHQD)(2)PHAL, employing various N-chlorinated hydantoins as the terminal chlorenium source. Ten examples are presented with selectivities ranging from 43 to 90% ee. This work represents the first example of an enantioselective reagent-controlled chlorolactonization that approaches synthetically useful enantioselectivities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245925
4-Pentenoic acid, 97%
C5H8O2
W284300
4-Pentenoic acid, ≥98%, stabilized, FG
C5H8O2