EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Citrinin selective molecularly imprinted polymers for SPE.
PMID 20175088
Abstract
Molecularly imprinted polymers (MIPs) for citrinin (Cit) with 1-hydroxy-2-naphthoic acid (HNA) as mimic template were prepared and the molecularly imprinted SPE method was developed for the detection of Cit in rice with HPLC. The adsorption properties of HNA and Cit on the MIPs and nonimprinted polymers were investigated. It proved that MIPs showed higher selectivity adsorption to HNA and Cit than nonimprinted polymers were. The recoveries of Cit in rice were in the range of 86.7-97.7%. The spiked rice samples and five rice samples in Beijing market were detected using molecularly imprinted SPE method and satisfied results were obtained as discussed in this article.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109630
1-Hydroxy-2-naphthoic acid, ≥97.0%
C11H8O3