EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Cholesterol trafficking is required for mTOR activation in endothelial cells.
PMID 20176935
Abstract
Mammalian target of rapamycin (mTOR) constitutes a nodal point of a signaling network that regulates cell growth and proliferation in response to various environmental cues ranging from growth factor stimulation to nutrients to stress. Whether mTOR is also affected by cholesterol homeostasis, however, has remained unknown. We report that blockade of cholesterol trafficking through lysosome by a newly identified inhibitor of angiogenesis, itraconazole, leads to inhibition of mTOR activity in endothelial cells. Inhibition of mTOR by itraconazole but not rapamycin can be partially restored by extracellular cholesterol delivered by cyclodextrin. Moreover, other known inhibitors of endosomal/lysosomal cholesterol trafficking as well as siRNA knockdown of Niemann-Pick disease type C (NPC) 1 and NPC2 also cause inhibition of mTOR in endothelial cells. In addition, both the accumulation of cholesterol in the lysosome and inhibition of mTOR caused by itraconazole can be reversed by thapsigarin. These observations suggest that mTOR is likely to be involved in sensing membrane sterol concentrations in endothelial cells, and the cholesterol trafficking pathway is a promising target for the discovery of inhibitors of angiogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U3633
U 18666A, powder
C25H41NO2·HCl