EMAIL THIS PAGE TO A FRIEND
Journal of Anesthesia
Sevoflurane postconditioning prevents activation of caspase 3 and 9 through antiapoptotic signaling after myocardial ischemia-reperfusion.
PMID 20177944