EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Endocrinology and Metabolism
In vivo exercise followed by in vitro contraction additively elevates subsequent insulin-stimulated glucose transport by rat skeletal muscle.
PMID 20179245
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-741
Anti-AS160 (Rab-GAP) Antibody, serum, clone, Upstate®