EMAIL THIS PAGE TO A FRIEND
Pancreas
Expression of transforming growth factor beta1/Sma- and Mad-related proteins in rat with chronic pancreatitis induced by dibutyltin dichloride.
PMID 20182310
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205494
Dibutyltin dichloride, 96%
C8H18Cl2Sn