EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Effect of nitroethane and nitroethanol on the production of indole and 3-methylindole (skatole) from bacteria in swine feces by gas chromatography.
PMID 20183070
Abstract
Indole and 3-methylindole (skatole) are odor pollutants in livestock waste, and skatole is a major component of boar taint. Skatole causes pulmonary edema and emphysema in ruminants and causes damage to lung Clara cells in animals and humans. A gas chromatographic method that originally used a nitrogen-phosphorus detector to increase sensitivity was modified resulting in an improved flame ionization detection response for indole and skatole of 236% and 207%, respectively. The improved method eliminates the large amount of indole decomposition in the injector. A 10 micro g mL(-1) spike of indole and skatole in water and swine fecal slurries resulted in recovery of 78.5% and 96% in water and 76.1% and 85.8% in fecal slurries, respectively. The effect of the addition of nitroethane and nitroethanol at 21.8 mM in swine fecal slurries was studied on the microbial production of indole and skatole. Nitroethane and nitroethanol decreased the production of skatole in swine fecal slurries at 24 h. The nitroethane effect on l-tryptophan-supplemented fecal slurries after 6 and 24 h incubation resulted in a decrease of 69.0% (P = 0.02) and 23.5% skatole production, respectively, and a decrease of 14.9% indole at 6 h, but an increase in indole production of 81.1% at 24 h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
227870
Nitroethane, ReagentPlus®, 99.5%
C2H5NO2
130206
Nitroethane, reagent grade, ≥98.0%
C2H5NO2