EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Structure and activity of carvedilol].
PMID 20184288
Abstract
Unique stereochemistry of the third-generation beta-adrenoblockers is a basis of advantages of the new drugs such as carvedilol. Data on the stereochemistry beta-adrenoblockers and the influence of their optical activity on the pharmacological effects of related drugs are presented. Main steps in the practical invest gation of carvedilol stereoisomers are reviewed. Clinical aspects of the use of carvedilol, representing a racemic mixture of two enantiomers, in the treatment chronic heart failure, arterial hypertension, and coronary heart disease are considered.