EMAIL THIS PAGE TO A FRIEND
Veterinary parasitology
In vivo evidence for the resistance of Theileria annulata to buparvaquone.
PMID 20185242
Abstract
The present study describes an outbreak of tropical theileriosis cases refractory to buparvaquone treatment, which occurred in a small-size dairy farm in Tunisia. Out of seven treated cows, four died in spite of repeated buparvaquone injections (2.5 and 5 mg kg(-1)) and the monitoring of the affected cows showed no improvement of the course of the disease with a consistent decrease in the haematocrit, the persistence of fever and an increased parasitaemia after treatment. Ticks were fed on a calf experimentally infected with one isolate established in culture from one of the cases and the resultant infected ticks ground up to generate a supernatant infected with the potentially resistant stock. This was used to experimentally infect three calves, and the clinical observations, post-buparvaquone treatment, showed an absence of the usual effect of buparvaquone treatment on the parasite Theileria annulata, such as the rapid decline of schizont index and parasitaemia and a rapid recovery from the disease. These results confirmed for the first time the occurrence of resistance to buparvaquone in the protozoan T. annulata.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32154
Buparvaquone, VETRANAL®, analytical standard
C21H26O3