EMAIL THIS PAGE TO A FRIEND
Huan jing ke xue= Huanjing kexue
[Study on the sorption of 4-octylphenol by freshwater algae].
PMID 20187402
Abstract
The sorption of 4-octylphenol (4-OP) by two freshwater algae was investigated. Results showed that the sorption of 4-octylphenol by algae was obvious and quick, where 20% of initial 4-OP (2 mg/L) was accumulated by Chlorella vulgaris (CV) and 46% initial 4-OP (2 mg/L) was accumulated by Anabaena cylindrical (AC) after 5 min incubation. The sorption got equilibrium at 1 h after incubation. Langmuir sorption model was good appropriate type for this sorption. The effect of pH value on CV sorption was obvious than that on AC sorption. The sorption capacity of the biomass of two algae increased with the decrease of pH value. The analyzing of interaction between algae and 4-octylphenol was performed by fluorescence spectrum. Results showed that the algae could weaker the fluorescence spectrum intensity of 4-octylphenol and result in red shift of the maximum absorbance wavelength of mixture solution. Based on the results, it was speculated that algae bound with the contamination could use the near UV region of solar radiation and induced the contamination degradation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
384445
4-Octylphenol, 99%
C14H22O
442850
4-Octylphenol, analytical standard
C14H22O