EMAIL THIS PAGE TO A FRIEND
International journal of antimicrobial agents
In vitro activity of biapenem against Burkholderia pseudomallei.
PMID 20188524
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0306
Biapenem, ≥98% (HPLC)
C15H18N4O4S