EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Stereochemistry-activity relationship of orally active tetralin S1P agonist prodrugs.
PMID 20188554
Abstract
Modifying FTY720, an immunosuppressant modulator, led to a new series of well phosphorylated tetralin analogs as potent S1P1 receptor agonists. The stereochemistry effect of tetralin ring was probed, and (-)-(R)-2-amino-2-((S)-6-octyl-1,2,3,4-tetrahydronaphthalen-2-yl)propan-1-ol was identified as a good SphK2 substrate and potent S1P1 agonist with good oral bioavailability.