EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and evaluation of tricyclic derivatives containing a non-aromatic amide as inhibitors of poly(ADP-ribose)polymerase-1 (PARP-1).
PMID 20189385
Abstract
Highly potent poly(ADP-ribose)polymerase-1 (PARP-1) inhibitors, including 9-hydroxy-1,2-dihydro-4H-thiopyrano[3,4-c]quinolin-5(6H)-one derivatives with a non-aromatic A-ring, were synthesized. Among the derivatives, 12a showed low nanomolar enzyme and cellular activity (IC(50) = 42 nM, ED(50) = 220 nM) with good water solubility. Further, 12a exhibited microsomal stability in vitro and brain permeability in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
159344
Ethyl 4-isocyanatobenzoate, 97%
C10H9NO3