EMAIL THIS PAGE TO A FRIEND
Molecular diversity
The Gewald multicomponent reaction.
PMID 20191319
Abstract
The Gewald reaction of sulfur, cyanoacetic acid derivatives, and oxo-component (G-3CR) yielding highly substituted 2-aminothiophene derivatives has seen diverse applications in combinatorial and medicinal chemistry. Its products are of great use in pharmaceutical industry mainly as small molecular weight inhibitors. We herein review synthetic scope and variations, usage, and structural biology of Gewald products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C88505
Cyanoacetic acid, 99%
C3H3NO2