EMAIL THIS PAGE TO A FRIEND
The American journal of psychiatry
Psychosis following excessive ingestion of energy drinks in a patient with schizophrenia.
PMID 20194494
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0100100 Haloperidol decanoate, European Pharmacopoeia (EP) Reference Standard
C31H41ClFNO3