EMAIL THIS PAGE TO A FRIEND
Blood
Putative GTPase GIMAP1 is critical for the development of mature B and T lymphocytes.
PMID 20194894
Abstract
The guanosine triphosphatases (GTPases) of the immunity-associated protein (GIMAP) family of putative GTPases has been implicated in the regulation of T-lymphocyte development and survival. A mouse conditional knockout allele was generated for the immune GTPase gene GIMAP1. Homozygous loss of this allele under the influence of the lymphoid-expressed hCD2-iCre recombinase transgene led to severe (> 85%) deficiency of mature T lymphocytes and, unexpectedly, of mature B lymphocytes. By contrast there was little effect of GIMAP1 deletion on immature lymphocytes in either B or T lineages, although in vitro studies showed a shortening of the survival time of both immature and mature CD4(+) single-positive thymocytes. These findings show a vital requirement for GIMAP1 in mature lymphocyte development/survival and draw attention to the nonredundant roles of members of the GIMAP GTPase family in these processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV50287
Anti-GIMAP1 antibody produced in rabbit, affinity isolated antibody