EMAIL THIS PAGE TO A FRIEND
Journal of economic entomology
Hydramethylnon uptake by Blattella germanica (Orthoptera: Blattellidae) by coprophagy.
PMID 2019680
Abstract
Blattella germanica (L.) that were fed hydramethylnon bait produced residues that were toxic to exposed conspecifics. Insecticidal activity was traced to the feces of treated insects by feeding radiolabeled material, where approximately 50% of the recovered radioactivity was unmetabolized parent compound. Ingestion of toxicant-laden feces by all life stages was evident, but the effect of this behavior was greatest on early instar nymphs. Baits containing toxicants with delayed activity, such as hydramethylnon, probably affect cockroach field populations indirectly through coprophagy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35373
Hydramethylnon, PESTANAL®, analytical standard
C25H24F6N4