EMAIL THIS PAGE TO A FRIEND
BMC medical genetics
Polymorphisms in IL-1beta, vitamin D receptor Fok1, and Toll-like receptor 2 are associated with extrapulmonary tuberculosis.
PMID 20196868
Abstract
Human genetic variants may affect tuberculosis susceptibility, but the immunologic correlates of the genetic variants identified are often unclear. We conducted a pilot case-control study to identify genetic variants associated with extrapulmonary tuberculosis in patients with previously characterized immune defects: low CD4+ lymphocytes and low unstimulated cytokine production. Two genetic association approaches were used: 1) variants previously associated with tuberculosis risk; 2) single nucleotide polymorphisms (SNPs) in candidate genes involved in tuberculosis pathogenesis. Single locus association tests and multifactor dimensionality reduction (MDR) assessed main effects and multi-locus interactions. There were 24 extrapulmonary tuberculosis cases (18 black), 24 pulmonary tuberculosis controls (19 black) and 57 PPD+ controls (49 black). In approach 1, 22 SNPs and 3 microsatellites were assessed. In single locus association tests, interleukin (IL)-1beta +3953 C/T was associated with extrapulmonary tuberculosis compared to PPD+ controls (P = 0.049). Among the sub-set of patients who were black, genotype frequencies of the vitamin D receptor (VDR) Fok1 A/G SNP were significantly different in extrapulmonary vs. pulmonary TB patients (P = 0.018). In MDR analysis, the toll-like receptor (TLR) 2 microsatellite had 76% prediction accuracy for extrapulmonary tuberculosis in blacks (P = 0.002). In approach 2, 613 SNPs in 26 genes were assessed. None were associated with extrapulmonary tuberculosis. In this pilot study among extrapulmonary tuberculosis patients with well-characterized immune defects, genetic variants in IL-1beta, VDR Fok1, and TLR2 were associated with an increased risk of extrapulmonary disease. Additional studies of the underlying mechanism of these genetic variants are warranted.