EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Biomimetic synthesis of metal ion-doped hierarchical crystals using a gel matrix: formation of cobalt-doped LiMn(2)O(4) with improved electrochemical properties through a cobalt-doped MnCO(3) precursor.
PMID 20198677
Abstract
We have synthesized spinel type cobalt-doped LiMn(2)O(4) (LiMn(2-y)Co(y)O(4), 0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
377449
Manganese(II) carbonate, ≥99.9% trace metals basis
CMnO3