EMAIL THIS PAGE TO A FRIEND
Neurology
Primary neuroprotection: the Holy Grail of multiple sclerosis therapy.
PMID 20200341
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0157
Ibudilast, ≥99% (HPLC), solid
C14 H18 N2 O