EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Comparison of two nitroxide labile esters for delivering electron paramagnetic resonance probes into mouse brain.
PMID 20201101
Abstract
In vivo quantitation of O(2) in brain has been hindered by a lack of suitable imaging modalities. Development of low-frequency electron paramagnetic resonance (EPR) spectrometers that can detect free radicals in animals in real time makes it feasible to image paramagnetic oximetry probes such as nitroxides in brain tissue. We have shown that masking the carboxyl group of 3-carboxy-2,2,5,5-tetramethyl-1-pyrrolidinyloxyl (nitroxide 1) as an esterase-labile acetoxymethyl ester yields 3-acetoxymethoxycarbonyl-2,2,5,5-tetramethyl-1-pyrrolidinyloxyl (nitroxide 2). Nitroxide 2 can cross the blood-brain barrier and is then hydrolyzed in situ by esterases to regenerate nitroxide 1, which becomes entrapped in brain tissue. Seeking to improve the loading of nitroxides into brain, we synthesized the more lipophilic pentanoyloxymethyl ester, 3-pentanoyloxymethoxycarbonyl-2,2,5,5-tetramethyl-1-pyrrolidinyloxyl (nitroxide 3). We report that the higher lipophilicity of nitroxide 3 does not significantly increase its ability to generate EPR signals in the mouse brain. Therefore, irrespective of whether nitroxide 2 or 3 was injected, similar levels of nitroxide were entrapped in brain tissue. These findings suggest that nitroxides 2 and 3 perform comparably well as proimaging agents for measuring O(2) distribution in brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253421
3-(2-Iodoacetamido)-PROXYL, free radical
C10H18IN2O2