EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Enantioselective organocatalytic addition of oxazolones to 1,1-bis(phenylsulfonyl)ethylene: a convenient asymmetric synthesis of quaternary alpha-amino acids.
PMID 20209524
Abstract
A new, easy, and highly enantioselective method for the synthesis of quaternary alpha-alkyl-alpha-amino acids based on organocatalysis is reported. The addition of oxazolones to 1,1-bis(phenylsulfonyl)ethylene is efficiently catalyzed by simple chiral bases or thioureas. The reaction affords alpha,alpha-disubstituted alpha-amino acid derivatives with complete C4 regioselectivity and with excellent yields and enantioselectivities. This methodology is complementary to previously reported enantioselective approaches to quaternary alpha-amino acids and allows the synthesis of alpha-phenyl-alpha-alkyl-alpha-amino acids and alpha-tert-butyl-alpha-alkyl-alpha-amino acids. It has distinct advantages in terms of operational simplicity, enviromentally friendly conditions, and suitability for large-scale reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15151
1,1-Bis(phenylsulfonyl)ethylene, ≥98.0% (CH)
C14H12O4S2