EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Total synthesis of (+)-chaetocin and its analogues: their histone methyltransferase G9a inhibitory activity.
PMID 20210309
Abstract
The first total synthesis of (+)-chaetocin has been accomplished in nine steps starting from known N-Cbz-N-Me-serine using radical alpha-bromination reaction of diketopiperazine 10 and Co(I)-mediated reductive dimerization reaction of 12 as key reactions. The enantiomers show comparable inhibitory activity toward histone methyltransferase (HMT) G9a, but analogues without the sulfur functionality are inactive.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9492
Chaetocin from Chaetomium minutum, ≥95% (HPLC)
C30H28N6O6S4