EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Complete nucleotide sequence and comparative analysis of pPR9, a 41.7-kilobase conjugative staphylococcal multiresistance plasmid conferring high-level mupirocin resistance.
PMID 20211895
Abstract
We have sequenced the conjugative plasmid pPR9, which carries the ileS2 gene, which had contributed to the dissemination of high-level mupirocin resistance at our institution. The plasmid backbone shows extensive genetic conservation with plasmids belonging to the pSK41/pGO1 family, but comparative analyses have revealed key differences that provide important insights into the evolution of these medically important plasmids and high-level mupirocin resistance in staphylococci and highlight the role of insertion sequence IS257 in these processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7694
Mupirocin, ≥92% (HPLC), powder
C26H44O9