EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Stable colloidal solutions of high-temperature-annealed L10 FePt nanoparticles.
PMID 20213650
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52270
1-Hexadecanethiol, ≥95.0% (GC)
C16H34S
674516
1-Hexadecanethiol, 99%
C16H34S