EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Organic thin-film transistors fabricated on resorbable biomaterial substrates.
PMID 20217767
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
754064
DH-FTTF, sublimed, 95%
C46H46S2