EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the chemical constituents of Portulaca oleracea].
PMID 20218290
Abstract
To study the chemical constituents of Portulaca oleracea. The constituents were isolated by column chromatography and identified on the basis of physicochemical and spectral data. Five compounds were isolated from 70% ethanol extract of this plant and their structures were elucidated as cyclo (Phe-Ile) (1), cycle (Tyr-Ala) (2), adenine (3), friedelin (4) and isoselachoceric acid (5). Compounds 1-5 are isolated from Portulaca oleracea for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
855022
Friedelin, technical grade
C30H50O
L6641
Lignoceric acid, ≥99% (GC)
C24H48O2