EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[GC-MS analysis of the liposoluble components from fruits of Harrisonia perforata].
PMID 20218293
Abstract
To extract and analyze the liposoluble chemical constituents of the petroleum ether extraction and alcohol extraction from fruits of Harrisonia perforata. To analyze the liposoluble components by GC-MS. 29 compounds were identified from petroleum ether extraction, amounting to 69.67% of the total detected constituents and the ir relative contents were confirmed. From the alcohol extraction, 12 compounds were identified, amounting to 71.00% of the total detected constituents and the ir relative contents were also confirmed. Linoleic acid, Palmitic acid, beta-Stitosterol are common liposoluble components in two extractions.