EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Cytoglobosins A-G, cytochalasans from a marine-derived endophytic fungus, Chaetomium globosum QEN-14.
PMID 20225834
Abstract
Cytoglobosins A-G (1-7), seven new cytochalasan derivatives, along with two structurally related known compounds, isochaetoglobosin D (8) and chaetoglobosin F(ex) (9), were isolated and identified from the cultures of Chaetomium globosum QEN-14, an endophytic fungus derived from the marine green alga Ulva pertusa. The structures of the new natural products as well as their relative configurations were elucidated on the basis of 1D and 2D NMR spectra (COSY, HSQC, HMBC, and NOESY) and HRESIMS data. Cytoglobosins C and D (3 and 4) displayed cytotoxic activity against the A-549 tumor cell line.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0642 Chaetoglobosin A, from Chaetomium globosum, ≥98% (HPLC)
C32H36N2O5