EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Gold nanoparticles for the improved anticancer drug delivery of the active component of oxaliplatin.
PMID 20225865
Abstract
The platinum-based anticancer drugs cisplatin, carboplatin, and oxaliplatin are an important component of chemotherapy but are limited by severe dose-limiting side effects and the ability of tumors to develop resistance rapidly. These drugs can be improved through the use of drug-delivery vehicles that are able to target cancers passively or actively. In this study, we have tethered the active component of the anticancer drug oxaliplatin to a gold nanoparticle for improved drug delivery. Naked gold nanoparticles were functionalized with a thiolated poly(ethylene glycol) (PEG) monolayer capped with a carboxylate group. [Pt(1R,2R-diaminocyclohexane)(H(2)O)(2)]2NO(3) was added to the PEG surface to yield a supramolecular complex with 280 (+/-20) drug molecules per nanoparticle. The platinum-tethered nanoparticles were examined for cytotoxicity, drug uptake, and localization in the A549 lung epithelial cancer cell line and the colon cancer cell lines HCT116, HCT15, HT29, and RKO. The platinum-tethered nanoparticles demonstrated as good as, or significantly better, cytotoxicity than oxaliplatin alone in all of the cell lines and an unusual ability to penetrate the nucleus in the lung cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
765708
Gold nanoparticles, 15 nm diameter, OD 50, methyl terminated, PEG 5000 coated, dispersion in H2O
765406
Gold nanoparticles, 50 nm diameter, amine functionalized, PEG 3000 coated, OD 50, dispersion in H2O
765449 Gold nanoparticles, 5 nm diameter, carboxylic acid functionalized, PEG 5000 coated, dispersion in H2O
765414
Gold nanoparticles, 50 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
765732
Gold nanoparticles, 30 nm diameter, methyl terminated, PEG 5000 coated, OD 50, dispersion in H2O
765317
Gold nanoparticles, 15 nm diameter, amine functionalized, PEG 3000 coated, OD 50, dispersion in H2O
765627
Gold nanoparticles, 10 nm diameter, methyl terminated, PEG 5000 coated, OD 50, dispersion in H2O
765694
Gold nanoparticles, 15 nm diameter, methyl terminated, PEG 2000 coated, OD 50, dispersion in H2O
765325
Gold nanoparticles, 15 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
753645
Gold nanoparticles, 50 nm diameter, OD 1, stabilized suspension in 0.1 mM PBS, reactant free
Au
765643
Gold nanoparticles, 40 nm diameter, methyl terminated, PEG 5000 coated, OD 50, dispersion in H2O
765309
Gold nanoparticles, 10 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
765686
Gold nanoparticles, 50 nm diameter, methyl terminated, PEG 5000 coated, OD 50, dispersion in H2O
765295
Gold nanoparticles, 10 nm diameter, amine functionalized, PEG 3000 coated, OD 50, dispersion in H2O
753688
Gold nanoparticles, 100 nm diameter, OD 1, stabilized suspension in 0.1 mM PBS, reactant free
Au
765341
Gold nanoparticles, 20 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
765724
Gold nanoparticles, 20 nm diameter, methyl terminated, PEG 5000 coated, OD 50, dispersion in H2O
765619
Gold nanoparticles, 10 nm diameter, methyl terminated, PEG 2000 coated, OD 50, dispersion in H2O
765368
Gold nanoparticles, 30 nm diameter, amine functionalize, PEG 3000 coated, OD 50, dispersion in H2O
765678
Gold nanoparticles, 50 nm diameter, methyl terminated, PEG 2000 coated, OD 50, dispersion in H2O
765279
Gold nanoparticles, 5 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
765333
Gold nanoparticles, 20 nm diameter, amine functionalized, PEG 3000 coated, OD 50, dispersion in H2O
765716
Gold nanoparticles, 20 nm diameter, methyl terminated, PEG 2000 coated, OD 50, dispersion in H2O
765384
Gold nanoparticles, 40 nm diameter, amine functionalized, PEG 3000 coated, OD 50, dispersion in H2O
765651
Gold nanoparticles, 40 nm diameter, methyl terminated, PEG 2000 coated, OD 50, dispersion in H2O
765392
Gold nanoparticles, 40 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
765635
Gold nanoparticles, 30 nm diameter, methyl terminated, PEG 2000 coated, OD 50, dispersion in H2O
765376
Gold nanoparticles, 30 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
765597
Gold nanoparticles, 5 nm diameter, methyl terminated, PEG 2000 coated, OD 50, dispersion in H2O
765503
Gold nanoparticles, 20 nm diameter, carboxylic acid functionalized, PEG 3000 coated, OD 50, dispersion in H2O