EMAIL THIS PAGE TO A FRIEND
Steroids
A practical Delta 1-dehydrogenation of Delta 4-3-keto-steroids with DDQ in the presence of TBDMSCl at room temperature.
PMID 20226803
Abstract
A mild and efficient Delta(1)-dehydrogenation of Delta(4)-3-keto-steroids with DDQ in the presence of tertbutyldimethylchlorosilane at room temperature was developed.