EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Oxidation of 2-t-butyl-4-methoxyphenol (BHA) by horseradish and mammalian peroxidase systems.
PMID 20227953
Abstract
Di-BHA, 2,2'-dihydroxy-3,3'-di-t-butyl-5,5'-dimethoxy-diphenyl, was isolated as the product of the reaction of either commercial horseradish peroxidase or partially purified rat intestine peroxidase (Donor-H(2)O(2) oxidoreductase, EC 1.11.1.7.) and hydrogen peroxide with 2-t-butyl-4-methoxyphenol (BHA). BHA, Di-BHA and other cyclic compounds possessing a hydroxyl group in the ring were found to be competitive inhibitors with respect to guaiacol, and non-competitive inhibitors with respect to hydrogen peroxide in a system containing guaiacol, hydrogen peroxide and peroxidase. A free radical intermediate generated during peroxidatic oxidation of BHA was detected and identified by means of EPR spectroscopy. It was estimated that during one hour incubation the peroxidase activity present in the rat ileum mucosa is able to oxidise 12mumoles BHA at a saturating concentration. It is suggested that peroxidative oxidation at the intestinal wall may represent a contribution to the inactivation of some phenol derivatives potentially toxic to mammals.