EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A visible dye-based staining method for DNA in polyacrylamide gels by ethyl violet.
PMID 20230772
Abstract
We describe a visible dye-based staining method for DNA in polyacrylamide gels using ethyl violet (EV). The novel method is a background-free, sensitive, economical, and simple procedure involving only staining and washing steps that can be completed within 30 min. As little as 0.8-1.6 ng of phiX174 DNA/HaeIII can be detected by EV, which is about eightfold more sensitive than Nile blue (NB) stain and twofold less sensitive than ethidium bromide (EB) stain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
228842
Ethyl Violet, cationic triarylmethane dye
C31H42ClN3