EMAIL THIS PAGE TO A FRIEND
International immunopharmacology
Ginsenoside Re enhances survival of human CD4+ T cells through regulation of autophagy.
PMID 20230918
Abstract
In the present study, we examined the effects of ginsenoside Re (Re) on cytokine expression, cytokine-dependent autophagy and cell survival in human CD4(+) T cells. When CD4(+) T cells isolated from human peripheral blood were treated with Re, LC3 and monodansylcadaverine (MDC), representative markers of autophagy, were decreased in a dose-dependent manner. Interestingly, Re suppressed the production of interferon-gamma (IFN-gamma) and immunity-related GTPase family M (IRGM) in CD4(+) T cells whereas no changes in other autophagy-related signaling molecules (ERK, p38 and AKT-mTOR-p70S6k) were found. Concomitantly, we observed that Re increased the proliferation of CD4(+) T cells with decreased cell death. Our results demonstrate that ginsenoside Re enhanced viability of CD4(+) T cells through the regulation of IFN-gamma-dependent autophagy activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4008
Dansylcadaverine, ≥97% (TLC)
C17H25N3O2S