EMAIL THIS PAGE TO A FRIEND
Annals of internal medicine
Summaries for patients. The effects of salsalate on blood sugar control in people with type 2 diabetes.
PMID 20231550
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0070
Salsalate, ≥98% (HPLC)
C14H10O5