EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Arctigenin blocks the unfolded protein response and shows therapeutic antitumor activity.
PMID 20232300
Abstract
Cancer cells in poorly vascularized solid tumors are constantly or intermittently exposed to stressful microenvironments, including glucose deprivation, hypoxia, and other forms of nutrient starvation. These tumor-specific conditions, especially glucose deprivation, activate a signaling pathway called the unfolded protein response (UPR), which enhances cell survival by induction of the stress proteins. We have established a screening method to discover anticancer agents that could preferentially inhibit tumor cell viability under glucose-deprived conditions. Here we identify arctigenin (ARC-G) as an active compound that shows selective cytotoxicity and inhibits the UPR during glucose deprivation. Indeed, ARC-G blocked expression of UPR target genes such as phosphorylated-PERK, ATF4, CHOP, and GRP78, which was accompanied by enhanced phosphorylation of eIF2 alpha during glucose deprivation. The UPR inhibition led to apoptosis involving a mitochondrial pathway by activation of caspase-9 and -3. Furthermore, ARC-G suppressed tumor growth of colon cancer HT-29 xenografts. Our results demonstrate that ARC-G can be served as a novel type of antitumor agent targeting the UPR in glucose-deprived solid tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00075
(−)-Arctigenin, ≥95% (LC/MS-ELSD)
C21H24O6