EMAIL THIS PAGE TO A FRIEND
Journal of the Chemical Society
Cinnolines; the preparation of 4-chloro-2-amino-acetophenone and related 4-hydroxycinnolines.
PMID 20238652
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
193968
4′-Chloro-3′-nitroacetophenone, 99%
C8H6ClNO3