EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
The estimation of urethane (ethylcarbamate) in blood.
PMID 20247716
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
517135
Diethyl carbonate, anhydrous, ≥99%
C5H10O3
D91551
Diethyl carbonate, 99%
C5H10O3