EMAIL THIS PAGE TO A FRIEND
Comptes rendus hebdomadaires des seances de l'Academie des sciences
[Not Available].
PMID 20248776
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
215112
Phosphorous acid, 99%
H3O3P
04115
Phosphorous acid solution, ≥50%
H3O3P