EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Use of sulfasuxidine, streptothricin, and streptomycin in nutritional studies with the chick.
PMID 20276107
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8752
Succinylsulfathiazole, ≥98%
C13H13N3O5S2